Anti-ISCA1 Mouse Monoclonal Antibody [clone: 2E9]

Supplier: Abnova
H00081689-M01
10592-946EA 502.5 CAD
10592-946
Anti-ISCA1 Mouse Monoclonal Antibody [clone: 2E9]
Antibodies
Type: Primary
Antigen: ISCA1
Clonality: Monoclonal
Clone: 2E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR