Anti-2610017G09Rik, ADP-GK Mouse Monoclonal Antibody [clone: 1E4]

Supplier: Abnova
H00083440-M01
10576-338EA 502.5 CAD
10576-338
Anti-2610017G09Rik, ADP-GK Mouse Monoclonal Antibody [clone: 1E4]
Antibodies
Type: Primary
Antigen: 2610017G09Rik, ADP-GK
Clonality: Monoclonal
Clone: 1E4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR