Anti-PMFBP1 Mouse Monoclonal Antibody [clone: 4G9]

Supplier: Abnova
H00083449-M01
10597-364EA 502.5 CAD
10597-364
Anti-PMFBP1 Mouse Monoclonal Antibody [clone: 4G9]
Antibodies
Type: Primary
Antigen: PMFBP1
Clonality: Monoclonal
Clone: 4G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR