Anti-AIF1L Mouse Polyclonal Antibody

H00083543-B01P
10552-218EA 420.99 CAD
10552-218
Anti-AIF1L Mouse Polyclonal Antibody
Type: Primary
Antigen: AIF1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR