Anti-AKR1E2 Mouse Polyclonal Antibody

Supplier: Abnova
H00083592-B01
10510-640EA 502.5 CAD
10510-640
Anti-AKR1E2 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: AKR1E2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR