Anti-MS4A8 Mouse Monoclonal Antibody [clone: 1B4]

Supplier: Abnova
H00083661-M04
10593-858EA 502.5 CAD
10593-858
Anti-MS4A8 Mouse Monoclonal Antibody [clone: 1B4]
Antibodies
Type: Primary
Antigen: MS4A8
Clonality: Monoclonal
Clone: 1B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR