Anti-SESN2 Mouse Monoclonal Antibody [clone: 3B8]

Supplier: Abnova
H00083667-M03
10601-124EA 502.5 CAD
10601-124
Anti-SESN2 Mouse Monoclonal Antibody [clone: 3B8]
Antibodies
Type: Primary
Antigen: SESN2
Clonality: Monoclonal
Clone: 3B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR