Anti-SLC25A18 Mouse Monoclonal Antibody [clone: 2F12]

Supplier: Abnova
H00083733-M02
10601-576EA 502.5 CAD
10601-576
Anti-SLC25A18 Mouse Monoclonal Antibody [clone: 2F12]
Antibodies
Type: Primary
Antigen: SLC25A18
Clonality: Monoclonal
Clone: 2F12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR