Anti-SETDB2 Mouse Monoclonal Antibody [clone: 100]

Supplier: Abnova
H00083852-M07
10570-998EA 502.5 CAD
10570-998
Anti-SETDB2 Mouse Monoclonal Antibody [clone: 100]
Antibodies
Type: Primary
Antigen: SETDB2
Clonality: Monoclonal
Clone: 100
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR