Anti-STK40 Mouse Monoclonal Antibody [clone: 4G3]

Supplier: Abnova
H00083931-M04
10602-774EA 502.5 CAD
10602-774
Anti-STK40 Mouse Monoclonal Antibody [clone: 4G3]
Antibodies
Type: Primary
Antigen: STK40
Clonality: Monoclonal
Clone: 4G3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR