Anti-EIF2A Mouse Monoclonal Antibody [clone: 3D5]

Supplier: Abnova
H00083939-M01
10584-792EA 502.5 CAD
10584-792
Anti-EIF2A Mouse Monoclonal Antibody [clone: 3D5]
Antibodies
Type: Primary
Antigen: EIF2A
Clonality: Monoclonal
Clone: 3D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR