Anti-TSSK6 Mouse Monoclonal Antibody [clone: 6F5]

Supplier: Abnova
H00083983-M02
10602-484EA 502.5 CAD
10602-484
Anti-TSSK6 Mouse Monoclonal Antibody [clone: 6F5]
Antibodies
Type: Primary
Antigen: TSSK6
Clonality: Monoclonal
Clone: 6F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR