Anti-FBXO30 Mouse Polyclonal Antibody

Supplier: Abnova
H00084085-A01
10556-438EA 364.5 CAD
10556-438
Anti-FBXO30 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FBXO30 (F-box protein 30)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR