Anti-ZIC4 Mouse Monoclonal Antibody [clone: 3G4]

Supplier: Abnova
H00084107-M12
10606-178EA 502.5 CAD
10606-178
Anti-ZIC4 Mouse Monoclonal Antibody [clone: 3G4]
Antibodies
Type: Primary
Antigen: ZIC4
Clonality: Monoclonal
Clone: 3G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR