Anti-LOXL4 Mouse Monoclonal Antibody [clone: 4H7]

Supplier: Abnova
H00084171-M01
10592-000EA 502.5 CAD
10592-000
Anti-LOXL4 Mouse Monoclonal Antibody [clone: 4H7]
Antibodies
Type: Primary
Antigen: LOXL4 (lysyl oxidase-like 4)
Clonality: Monoclonal
Clone: 4H7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR