Anti-SLA2 Mouse Monoclonal Antibody [clone: 3G5]

Supplier: Abnova
H00084174-M01
10601-466EA 502.5 CAD
10601-466
Anti-SLA2 Mouse Monoclonal Antibody [clone: 3G5]
Antibodies
Type: Primary
Antigen: SLA2
Clonality: Monoclonal
Clone: 3G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR