Anti-MEX3B Mouse Monoclonal Antibody [clone: 4C4]

Supplier: Abnova
H00084206-M05
10599-828EA 502.5 CAD
10599-828
Anti-MEX3B Mouse Monoclonal Antibody [clone: 4C4]
Antibodies
Type: Primary
Antigen: MEX3B
Clonality: Monoclonal
Clone: 4C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR