Anti-CAMKK1 Mouse Monoclonal Antibody [clone: 3B4]

Supplier: Abnova
H00084254-M05
10578-570EA 502.5 CAD
10578-570
Anti-CAMKK1 Mouse Monoclonal Antibody [clone: 3B4]
Antibodies
Type: Primary
Antigen: CAMKK1
Clonality: Monoclonal
Clone: 3B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR