Anti-FBXW9 Mouse Polyclonal Antibody

Supplier: Abnova
H00084261-B01
10556-170EA 502.5 CAD
10556-170
Anti-FBXW9 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FBXW9 (F-box and WD repeat domain containing 9)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR