Anti-PDCD2L Mouse Monoclonal Antibody [clone: 5F9]

Supplier: Abnova
H00084306-M01
10593-326EA 502.5 CAD
10593-326
Anti-PDCD2L Mouse Monoclonal Antibody [clone: 5F9]
Antibodies
Type: Primary
Antigen: PDCD2L
Clonality: Monoclonal
Clone: 5F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR