Anti-SARNP Mouse Polyclonal Antibody

Supplier: Abnova
H00084324-B01
10553-534EA 502.5 CAD
10553-534
Anti-SARNP Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SARNP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR