Valsartan
Synonyms:
(S)-2-(N-((2'-(1H-Tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)pentanamido)-3-methylbutanoic acid
, (S)-2-(N-((2'-(1H-Tetrazol-5-yl)[1,1'-biphenyl]-4-yl)methyl)pentanamido)-3-methylbutanoic acid
2004-500
2004-25
2004-100
10188-686EA
614.54
CAD
10188-686
10188-688
10188-684
Valsartan
Valsartan
Formula:
C₂₄H₂₉N₅O₃ MW: 435.52 g/mol |
MDL Number:
MFCD00865840 CAS Number: 137862-53-4 |
Specification Test Results
CAS | 137862-53-4 |
Purity | ≥98% by TLC |
Form | White solid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...