Sulfo-NHS (N-Hydroxysulfosuccinimide sodium salt)

Supplier: BioVision

Synonyms: Sulpho-NHSHydroxy-2,5-dioxopyrrolidine-3-sulfonicacid sodium saltSulfo-NHSHydroxy-2,5-dioxopyrrolidine-3-sulphonicacid sodium saltSulpho-N-HydroxysuccinimideSulfo-N-Hydroxysuccinimide

2321-1EA 2321-5G 2321-500
10189-566EA 2683.95 CAD
10189-566 10189-564 10189-568
Sulfo-NHS (N-Hydroxysulfosuccinimide sodium salt)
N-Hydroxysulfosuccinimide sodium salt (Sulfo-NHS)
Formula: C₄H₄NNaO₆S
MW: 217.13 g/mol
MDL Number: MFCD00043100
CAS Number: 106627-54-7

Order Now

Specification Test Results

CAS 106627-54-7
Purity ≥95% by NMR
Form White to off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR