Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-C20ORF24 (chromosome 20 open reading frame 24) Rabbit Polyclonal Antibody

Supplier: Genetex

Rabbit polyclonal antibody to C20ORF24 (N-terminal)

Recommended Dilutions: For Western Blot: Use at 2.5 ug/ml.<p>Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: C20ORF24 (chromosome 20 open reading frame 24)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μg GTX46085 89288-430 Each (100µG) Retrieving Restricted
restricted You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antibody type Primary
Antigen name Chromosome 20 Open Reading Frame 24
Antigen symbol C20ORF24 (chromosome 20 open reading frame 24)
Antigen synonyms PNAS-11, RIP5
Clonality Polyclonal
Concentration 1 mg/ml
Conjugation Unconjugated
Gene ID 55969
Host Rabbit
Immunogen Synthetic peptide
Isotype IgG
Purification Protein A Purified Total IgG
Reactivity Human, Rat, Mouse
Storage Buffer Phosphate buffered saline (PBS)
Storage temperature Store at 4°C for short-term use. For longer periods of storage, aliquot and store at -20°C. Avoid repeat freeze-thaw cycles.
Western Blot Yes
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty