Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD2 Rabbit Monoclonal Antibody [clone: EPR3693]

Supplier: Genetex
GTX62240
Rabbit monoclonal antibody [EPR3693] to CD2

Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: EPR3693
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μl GTX62240 89326-702 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol CD2
Antigen name CD2 molecule
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone EPR3693
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoFluorescence Yes
Antigen synonyms T11, T-cell surface antigen T11, LFA-3 receptor, LFA-2, erythrocyte receptor, Leu-5, SRBC, rosette receptor, sheep red blood cell receptor, lymphocyte-function antigen-2
Gene ID 914
Immunogen Synthetic peptide corresponding to residues in human CD2.
Purification Whole antiserum
Storage buffer pH: 7.2, Preservative: 0.01% Sodium azide. Constituents: 50% Glycerol, 0.05% BSA
Storage temperature Store at -20°C. Stable for 12 months at -20°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty