Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TMPRSS2 Rabbit Monoclonal Antibody [clone: EPR3862]

Supplier: Genetex
Rabbit monoclonal antibody [EPR3862] to TMPRSS2

IHC-P: 1/1000 - 1/2000. Perform heat mediated antigen retrieval before commencing with IHC staining protocol. ICC/IF: 1/150. WB: 1/1000 - 1/5000. Detects a band of approximately 54,28 kDa.

Type: Primary
Antigen: TMPRSS2
Clonality: Monoclonal
Clone: EPR3862
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μl GTX62636 89327-302 Each (100µl) Retrieving Restricted
restricted You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol TMPRSS2
Antigen name Transmembrane Protease Serine 2
Reactivity Rat, Mouse, Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone EPR3862
Host Rabbit
Isotype IgG
Western Blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Antigen synonyms Transmembrane protease serine 2, Transmembrane protease serine 2 catalytic chain, Transmembrane protease serine 2 non-catalytic chain, Serine protease 10, TMPRSS2, PRSS10
Gene ID 7113
Immunogen the immunogen in the range of amino acids 460-492 a.a of human TMPRSS2
Purification Protein A purified
Storage Buffer 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol, 0.01% sodium azide and 0.05% BSA.
Storage temperature Store at -20 °C. Stable for 12 months from date of receipt.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty