Anti-PLA2G7 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA526819
CAPIPA526819EA 3084.29 CAD
CAPIPA526819
Anti-PLA2G7 Rabbit Polyclonal Antibody
Antibodies
The protein encoded by this gene is a secreted enzyme that catalyzes the degradation of platelet-activating factor to biologically inactive products. Defects in this gene are a cause of platelet-activating factor acetylhydrolase deficiency.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:10-1:50, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: PLA2G7 (phospholipase A2, group VII (platelet-activating factor acetylhydrolase, plasma))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR