Anti-NEDD4 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA526930
CAPIPA526930EA 3025.12 CAD
CAPIPA526930
Anti-NEDD4 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with rat based on sequence homology. Nedd4 is one of a group of mouse genes that show developmentally regulated expression in mouse embryonic brain. Nedd4 is expressed in various other embryonic tissues and persists in most adult tissues.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: NEDD4 (neural precursor cell expressed, developmentally down-regulated 4, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR