Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VAMP7 Rabbit Polyclonal Antibody

Supplier: Prosci
6929_S
VAMP7 Antibody: VAMP7 is a member of the soluble N-ethylmaleimide-sensitive factor attachment protein receptor (SNARE) family, localizing to late endosomes and lysosomes. VAMP7 is thought to mediate the fusion of endosomes to their target lysosomes as well as other exocytosis events during phagocytosis and neuritogenesis. VAMP7 interacts with the VPS9 ankyrin repeat protein VARP, a protein that localizes to early endosomes and thought to regulate endosome dynamics. Together with CD82, VAMP7 can modulate the signaling of EGFR by regulating its endocytosis from the plasma membrane.

Type: Primary
Antigen: VAMP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-Terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
20 µg 6929_S 10751-680 Each (20µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol VAMP7
Antigen name Vesicle-Associated Membrane Protein 7
Reactivity Mouse, Rat
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoFluorescence Yes
Antigen synonyms Vesicle-associated membrane protein 7, SYBL1, Synaptobrevin-like protein 1, Tetanus-insensitive VAMP, Ti-VAMP, VAMP-7, VAMP7
Epitope near the N-Terminus
Concentration 1 mg/ml
Cross adsorption No
Immunogen VAMP7 antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human VAMP7.
Purification affinity chromatography purified via peptide column.
Form Liquid
Storage buffer PBS containing 0.02% sodium azide.
Storage temperature VAMP7 antibody can be stored at 4°C for three months and -20°C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Shipping temperature -20°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty