Order Entry
ContactUsLinkComponent
Anti-PHLPP2 Rabbit Polyclonal Antibody
Anti-PHLPP2 Rabbit Polyclonal Antibody
Catalog #: 10751-738
Supplier:  Prosci
Anti-PHLPP2 Rabbit Polyclonal Antibody
Catalog #: 10751-738
Supplier:  Prosci
Price
292,68 $
Log in for availability
Unit of Measure Each (20µG)
Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    PH domain and leucine rich repeat protein phosphatase 2
  • Clonality:
    Polyclonal
  • Gene ID:
    Q6ZVD8
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Antigen symbol:
    PHLPP2
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • ImmunoFluorescence:
    Yes
  • Size:
    0.02 mg
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Antigen synonyms:
    PHLPP2
  • Storage buffer:
    PHLPP2 Antibody is supplied in PBS containing 0.02% sodium azide.
  • Molecular weight:
    156 kDa
  • Storage temperature:
    PHLPP2 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
  • Concentration:
    1 mg/mL
  • Shipping temperature:
    4 °C
  • Immunogen:
    PHLPP2 antibody was raised against a 20 amino acid synthetic peptide near the carboxy terminus of human PHLPP2. The immunogen is located within the last 50 amino acids of PHLPP2.
  • Tested applications:
    Immunocytochemistry
  • Purification:
    PHLPP2 Antibody is affinity chromatography purified via peptide column.
  • Cat. no.:
    10751-738

Specifications

About this item

PHLPP2 Antibody: PHLPP2 is a member of the serine/threonine phosphatase family, which are important regulators of Akt serine-threonine kinases (AKT1, AKT2, AKT3) and conventional/novel protein kinase C (PKC) isoforms. PHLPP1 and PHLPP2 have a similar domain structure and have been shown to dephosphorylate and inactivate, distinct Akt isoforms, at one of the two critical phosphorylation sites required for activation: Serine473. PHLPP2 dephosphorylates AKT1 and AKT3, whereas PHLPP1 is specific for AKT2 and AKT3. PHLPP1 promotes apoptosis and may act as a tumor suppressor. PHLPP2 associates with and is inhibited by adenylyl cyclase type 6 (AC6), thereby allowing Akt activation.

Type: Primary
Antigen: PHLPP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Customers Who Bought This Also Bought