Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MYCT1 Rabbit Polyclonal Antibody

Supplier: Prosci
7031_S
MYCT1 Antibody: MYCT1 was initially identified as a novel target of the c-Myc oncogene in myeloid cells. It is a widely expressed nuclear protein whose overexpression can promote apoptosis, alteration of morphology, enhancement of anchorage-independent cell growth, tumorigenic conversion, promotion of genomic instability, and inhibition of hematopoietic differentiation. MYCT1 binds to the promoters of several c-Myc-regulated genes and it has been suggested that the phenotypes seen in MYCT1-overexpressing cells are a result of the deregulation of these genes. RUNX1-ETO, a fusion protein made up of RUNX1 and ETO, is thought to deregulate the proliferation and responsiveness of human hematopoietic progenitor cells downstream of MYCT1.

Type: Primary
Antigen: MYCT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
20 µg 7031_S 10751-748 Each (20µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol MYCT1
Antigen name Myc Target In Myeloid Cells 1
Reactivity Mouse, Rat, Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Antigen synonyms MTLC, MYCT1, Myc target protein 1, Myc target in myeloid cells protein 1, MTMC1
Concentration 1 mg/ml
Cross adsorption No
Immunogen MYCT1 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human MYCT1.
Purification affinity chromatography purified via peptide column.
Form Liquid
Storage buffer PBS containing 0.02% sodium azide.
Storage temperature MYCT1 antibody can be stored at 4°C for three months and -20°C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Shipping temperature -20°C

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty