Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VNN3 Rabbit Polyclonal Antibody

Supplier: Prosci
7057_S
VNN3 Antibody: The vanin family is a novel group of ectoenzymes that function in tissue repair and plays a role in oxidative-stress response. As both secreted and membrane proteins, the vanin family members have been implicated as therapeutic targets in inflammatory disease. VNN3 (vascular non-inflammatory molecule 3), also known as Vanin3, is a 501 amino acid GPI-anchored amidohydrolase that is widely expressed and is found at highest levels in blood and liver. Induced by Th17 / Th1 type cytokines, VNN3 converts pantetheine into pantothenic acid. Containing one CN hydrolase domain, VNN3 is encoded by a gene that maps to human chromosome 6q23.2.

Type: Primary
Antigen: VNN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
20 µg 7057_S 10751-770 Each (20µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol VNN3
Antigen name Vanin 3
Reactivity Human, Mouse, Rat
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Antigen synonyms Vanin-3, VNN3, Vascular non-inflammatory molecule 3, HSA238982
Concentration 1 mg/ml
Cross adsorption No
Immunogen VNN3 antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human VNN3.
Purification affinity chromatography purified via peptide column.
Form Liquid
Storage buffer PBS containing 0.02% sodium azide.
Storage temperature VNN3 antibody can be stored at 4°C for three months and -20°C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Shipping temperature -20°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty