Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-POU3F2 Rabbit Polyclonal Antibody

Supplier: Prosci
7085_S
POU3F2 Antibody: POU3F2, also commonly known as brain-2, is a member of a family of POU domain genes expressed in mouse brain and is thought to be involved in the development of the neocortex and establishment of neural cell lineage. Recent studies suggest that POU3F2 may be involved with the development of neurodegenerative diseases as well as tumor development and proliferation. Along with the neural-lineage-specific transcription factors ASCL1 and MYT1L, POU3F2 can convert fibroblasts to functional neurons in vitro, a form of artificial stem cells termed induced neuronal (iN) cells, suggesting that these cells may be useful in the treatment of neurodegenerative diseases.

Type: Primary
Antigen: POU3F2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
20 µg 7085_S 10751-792 Each (20µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol POU3F2
Antigen name POU Class 3 Homeobox 2
Reactivity Rat, Human, Mouse
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoFluorescence Yes
Antigen synonyms brn-2, N-Oct3, OTF7, OTF-7, BRN2, POUF3, OCT7, oct-7
Concentration 1 mg/ml
Cross adsorption No
Immunogen Rabbit polyclonal POU3F2 antibody was raised against an 18 amino acid peptide near the carboxy terminus of human POU3F2.
Purification affinity chromatography purified via peptide column.
Form Liquid
Storage buffer PBS containing 0.02% sodium azide.
Storage temperature POU3F2 antibody can be stored at 4°C for three months and -20°C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Shipping temperature -20°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty