Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HEPACAM2 Rabbit Polyclonal Antibody

Supplier: Prosci
7111_S
HEPACAM2 Antibody: HEPACAM2 (Hepatocyte cell adhesion molecule 2), a type I N-linked transmembrane glycoprotein, belongs to the immunoglobulin superfamily. The exact function of HEPACAM2 is currently unknown, but the related protein HEPACAM forms cishomodimers on the cell surface to regulate the cell adhesion and may inhibit cell growth through suppression of cell proliferation. HEPACAM and HEPACAM2 mRNA are differentially regulated in canine tumors, with HEPACAM2 mRNA showing increased levels in adenomas, but decreased levels in metastatic carcinomas compared to normal tissues, while HEPACAM protein levels decreased in adenomas. It is therefore likely that HEPACAM2 plays a different role than HEPACAM in the development and progression to tumors.

Type: Primary
Antigen: HEPACAM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
20 µg 7111_S 10751-814 Each (20µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol HEPACAM2
Reactivity Mouse, Rat, Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Antigen synonyms UNQ305/PRO346, HEPACAM family member 2, Mitotic kinetics regulator, HEPACAM2, MIKI
Concentration 1 mg/ml
Cross adsorption No
Immunogen Rabbit polyclonal HEPACAM2 antibody was raised against a 19 amino acid peptide near the carboxy terminus of human HEPACAM2.
Purification affinity chromatography purified via peptide column.
Form Liquid
Storage buffer PBS containing 0.02% sodium azide.
Storage temperature HEPACAM2 antibody can be stored at 4°C for three months and -20°C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Shipping temperature -20°C

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty