Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RTN4RL2 Rabbit Polyclonal Antibody

Supplier: Prosci
7145_S
RTN4RL2 Antibody: Reticulon 4 receptor-like 2 (RTN4RL2), also known as NgR2and NgRH1, is a 60 kDa glycosylphosphatidylinositol-anchored protein that is expressed primarily in the brain. It contains eight leucine-rich repeats that are flanked by cysteine-rich sequences at both the N- and C-termini. The membrane anchored RTN4RL2 can be solubilized through the action of phospholipase or an unidentified MTMMP to generate a 46 kDa soluble receptor. It has been suggested that RTN4RL2 may play a role in regulating axonal regeneration and plasticity in the adult central nervous system.

Type: Primary
Antigen: RTN4RL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
20 µg 7145_S 10751-828 Each (20µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol RTN4RL2
Antigen name Reticulon 4 Receptor-like 2
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Antigen synonyms Reticulon-4 receptor-like 2, NGRL3, NGRH1, RTN4RL2, NgR2, Nogo receptor-like 3
Concentration 1 mg/ml
Cross adsorption No
Immunogen Rabbit polyclonal RTN4RL2 antibody was raised against an 18 amino acid peptide near the carboxy terminus of human RTN4RL2.
Purification affinity chromatography purified via peptide column.
Form Liquid
Storage buffer PBS containing 0.02% sodium azide.
Storage temperature RTN4RL2 antibody can be stored at 4°C for three months and -20°C, stable for up to one year.
Shipping temperature -20°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty