Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD5 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 53-7.3]

Supplier: Biolegend
100612
Alexa Fluor® 488 anti-mouse CD5 [53-7.3]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IF, IHC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 10<sup>6</sup> cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.<BR><BR>* Alexa Fluor® 488 has a maximum emission of 519 nm when it is excited at 488 nm.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: 53-7.3
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg 100612 CA100612-BL Each (100µG) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol CD5
Antigen name CD5 molecule
Reactivity Mouse
Conjugation Alexa Fluor® 488
Clonality Monoclonal
Antibody type Primary
Clone
Host Rat
Isotype IgG2a kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Antigen synonyms T-cell surface glycoprotein CD5, T1, LEU1, CD5, Lymphocyte antigen T1/Leu-1
Gene ID
Concentration
Immunogen
Purification
Form
Storage temperature
Shipping temperature
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty