Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD54 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: YN1/1.7.4]

Supplier: Biolegend
116111 116112
Alexa Fluor® 488 anti-mouse CD54 [YN1/1.7.4]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, IF, IHC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD54
Clonality: Monoclonal
Clone: YN1/1.7.4
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg 116112 CA116112-BL Each (100µG) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

25 µg 116111 CA116111-BL Each (25µG) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol ICAM1
Antigen name Intercellular Adhesion Molecule 1
Reactivity Mouse
Conjugation Alexa Fluor® 488
Clonality Monoclonal
Antibody type Primary
Clone
Host Rat
Isotype IgG2b kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Antigen synonyms intercellular adhesion molecule 1 (CD54), P3.58, human rhinovirus receptor, CD54, BB2, major group rhinovirus receptor, ICAM-1 BB2, major group rhinovirus receptor, ICAM-1, P3.58, CD54, human rhinovirus receptor, intercellular adhesion molecule 1 (CD54)
Gene ID
Concentration
Immunogen
Purification
Form
Storage temperature
Shipping temperature
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping