Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD69 Armenian Hamster Monoclonal Antibody (Alexa Fluor® 488) [clone: H1.2F3]

Supplier: Biolegend
104516
Alexa Fluor® 488 anti-mouse CD69 [H1.2F3]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC, IF; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: H1.2F3
Conjugation: Alexa Fluor® 488
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg 104516 CA104516-BL Each (100µG) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol CD69
Antigen name CD69 molecule
Reactivity Mouse
Conjugation Alexa Fluor® 488
Clonality Monoclonal
Antibody type Primary
Clone
Host Hamster
Isotype IgG
ImmunoChemistry Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Antigen synonyms CLEC2C, BL-AC/P26, EA1, AIM, GP32/28, MLR-3
Gene ID
Concentration
Immunogen
Purification
Form
Storage temperature
Shipping temperature
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty