Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD90.2 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 30-H12]

Supplier: Biolegend
105316 105315
Alexa Fluor® 488 anti-mouse CD90.2 [30-H12]; Isotype: Rat IgG2b, κ; Reactivity: Mouse, Does not react with Thy-1.1 (CD90.1); Apps: FC, IHC-F, IF; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. For immunohistochemical staining on frozen tissue sections, a concentration range of 5 - 10 μg/ml is suggested. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD90.2
Clonality: Monoclonal
Clone: 30-H12
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse;Does not react with Thy-1.1 (CD90.1)




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg 105316 CA105316-BL Each (100µG) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

25 µg 105315 CA105315-BL Each (25µG) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol CD90.2
Antigen name CD90.2 Protein
Reactivity Mouse
Conjugation Alexa Fluor® 488
Clonality Monoclonal
Antibody type Primary
Clone
Host Rat
Isotype IgG2b kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Antigen synonyms Thymus cell Antigen 1 Theta
Gene ID
Concentration
Immunogen
Purification
Form
Storage temperature
Shipping temperature
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty