Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-H-2Kb Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: AF6-88.5]

Supplier: Biolegend
116510
Alexa Fluor® 488 anti-mouse H-2Kb [AF6-88.5]; Isotype: Mouse (BALB/c) IgG2a, κ; Reactivity: Mouse b haplotype, Does not cross-react with other haplotypes (e.g., d, f, j, k, p, q, r, s, u, v); Apps: FC, IF, IHC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: H-2Kb
Clonality: Monoclonal
Clone: AF6-88.5
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: Mouse (BALB/c) IgG2a, κ
Reactivity: Mouse b haplotype;Does not cross-react with other haplotypes (e.g.;d;f;j;k;p;q;r;s;u;v)




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg 116510 CA116510-BL Each (100µG) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol H-2Kb
Antigen name MHC Class I (H-2Kb)
Reactivity Mouse
Conjugation Alexa Fluor® 488
Clonality Monoclonal
Antibody type Primary
Clone
Host Mouse
Isotype IgG2a kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Antigen synonyms K-Q alpha chain, MHC class I heavy chain H2-K, H2-K, MHC class I H2-K-b-alpha-2 cell surface glycoprotein, H-2K(d), H-2 class I histocompatibility antigen, H-2K, MHC H2-K transplantation antigen, H-2K(K), K-f,H-2 class I histocompatibility antigen, K-B alpha chain, class I major histocom, K-D alpha chain, H-2K(Q), H-2K(B), MHC class I antigen, K-K alpha chain, K-W28 alpha chain
Gene ID
Concentration
Immunogen
Purification
Form
Storage temperature
Shipping temperature
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty