Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-H-2Kd Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: SF1-1.1]

Supplier: Biolegend
116609 116610
Alexa Fluor® 488 anti-mouse H-2Kd [SF1-1.1]; Isotype: Mouse (SJL) IgG2a, κ; Reactivity: Mouse d haplotype, Weakly cross-reacts with H-2k haplotype, Does not cross react with other haplotypes (e.g., b, j, p, q, s, v).; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: H-2Kd
Clonality: Monoclonal
Clone: SF1-1.1
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: Mouse (SJL) IgG2a, κ
Reactivity: Mouse d haplotype;Weakly cross-reacts with H-2k haplotype;Does not cross react with other haplotypes (e.g.;b;j;p;q;s;v).




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg 116610 CA116610-BL Each (100µG) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

25 µg 116609 CA116609-BL Each (25µG) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol H-2KD
Antigen name MHC Class I (H-2Kd)
Reactivity Mouse
Conjugation Alexa Fluor® 488
Clonality Monoclonal
Antibody type Primary
Clone
Host Mouse
Isotype IgG2a kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Antigen synonyms H2DD, K-D alpha chain, H2K1, H2KD, H-2 class I histocompatibility antigen K-D alpha chain, H2KD, H-2 class I histocompatibility antigen, H2K1, H2DD
Gene ID
Concentration
Immunogen
Purification
Form
Storage temperature
Shipping temperature
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping