Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-I-A/I-E Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: M5/114.15.2]

Supplier: Biolegend
107616 107615
Alexa Fluor® 488 anti-mouse I-A/I-E [M5/114.15.2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, IF, IHC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: I-A/I-E
Clonality: Monoclonal
Clone: M5/114.15.2
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg 107616 CA107616-BL Each (100µG) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

25 µg 107615 CA107615-BL Each (25µG) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol I-A/I-E
Antigen name
Reactivity Mouse
Conjugation Alexa Fluor® 488
Clonality Monoclonal
Antibody type Primary
Clone
Host Rat
Isotype IgG2b kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Antigen synonyms
Gene ID
Concentration
Immunogen
Purification
Form
Storage temperature
Shipping temperature

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping