Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NK-1.1 Mouse Monoclonal Antibody (Alexa Fluor® 488)

Supplier: Biolegend
108717 108718
Type: Primary
Antigen: NK-1.1
Clonality: Monoclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg 108718 CA108718-BL Each (100µG) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

25 µg 108717 CA108717-BL Each (25µG) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol NK1.1
Antigen name NK-1.1 Antigen
Reactivity Mouse
Conjugation Alexa Fluor® 488
Clonality Monoclonal
Antibody type Primary
Host Mouse
Isotype IgG2a kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Antigen synonyms natural killer cell surface protein P1-40, ly-55c,CD161 antigen-like family member C, Ly59, NK1.1, natural killer cell receptor-P1, Nk-1, NKR-P1.9, NK-RP1, AI462337, natural killer cell-associated antigen 1, locus C, NKRP1, NKR-P1 40, natural killer cell receptor protein NKR-P1C, Nkrp1c, lymphocyte antigen 55c, killer cell lectin-like receptor subfamily A member 1C, Nk-1.2, lymphocyte antigen 55 complex, CD161, lymphocyte antigen 59, Nk1, Ly55c, Ly-59, Nk1.2, NKRP140 Nk1.2, Ly59, NKRP140, natural killer cell-associated antigen 1, NK-RP1, Nk-1, locus C, NKR-P1 40, natural killer cell surface protein P1-40, NKRP1, ly-55c,CD161 antigen-like family member C, CD161, NK1.1, AI462337, Ly-59, Nk-1.2, killer cell lectin-like receptor subfamily A member 1C, lymphocyte antigen 55 complex, NKR-P1.9, natural killer cell receptor protein NKR-P1C, lymphocyte antigen 55c, Ly55c, Nkrp1c, lymphocyte antigen 59, Nk1, natural killer cell receptor-P1
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty