Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD44 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: IM7]

Supplier: Biolegend
103016 103015
Alexa Fluor® 488 anti-mouse/human CD44 [IM7]; Isotype: Rat IgG2b, κ; Reactivity: Mouse, Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus, Squirrel Monkey, Horse (Equine), Cattle (Bovine, Cow), Swine (Pig, Porcine), Dog (Canine), Cat (Feline); Apps: FC, IF, IHC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: IM7
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse;Human;Cross-Reactivity: Chimpanzee;Baboon;Cynomolgus;Rhesus;Squirrel Monkey;Horse (Equine);Cattle (Bovine;Cow);Swine (Pig;Porcine);Dog (Canine);Cat (Feline)




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg 103016 CA103016-BL Each (100µG) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

25 µg 103015 CA103015-BL Each (25µG) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol CD44
Antigen name Cluster of Differentiation 44 antigen
Reactivity Rhesus Monkey, Cynomolgus Monkey, Mouse, Baboon, Bovine, Human, Dog, Pig, Cat Pig, Dog, Mouse, Baboon, Bovine, Cat, Cynomolgus Monkey, Human, Rhesus Monkey
Conjugation Alexa Fluor® 488
Clonality Monoclonal
Antibody type Primary
Clone
Host Rat
Isotype IgG2b kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Antigen synonyms MC56, MIC4, LHR, Pgp1, ECMR-III, HCELL, HCAM, MDU3, CSPG8, IN, CDW44, HUTCH-I, MDU2 MDU3, MIC4, CSPG8, HCAM, HUTCH-I, Pgp1, ECMR-III, IN, CDW44, LHR, MC56, MDU2, HCELL
Gene ID
Concentration
Immunogen
Purification
Form
Storage temperature
Shipping temperature
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty