Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD25 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: OX-39]

Supplier: Biolegend
Alexa Fluor® 488 anti-rat CD25 [OX-39]; Isotype: Mouse IgG1, κ; Reactivity: Rat; Apps: FC, IHC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD25
Clonality: Monoclonal
Clone: OX-39
Conjugation: Alexa Fluor® 488
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Rat

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg 202108 CA202108-BL Each (100µG) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol IL2RA
Antigen name Interleukin 2 Receptor, Alpha
Reactivity Rat
Conjugation Alexa Fluor® 488
Clonality Monoclonal
Antibody type Primary
Host Mouse
Isotype IgG1 kappa
ImmunoChemistry Yes
Flow cytometry Yes
Antigen synonyms IL-2 receptor subunit alpha, IL2R, TCGFR, p55, IL-2-RA, CD25, IL2-RA, TAC antigen, IL-2R subunit alpha, IDDM10
Gene ID
Storage temperature
Shipping temperature
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty