Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Tubulin-alpha Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 10D8]

Supplier: Biolegend
627905 627906
Alexa Fluor® 488 anti-Tubulin-α [10D8]; Isotype: Mouse IgM, κ; Reactivity: Human, Mouse, Rat, All Species; Apps: IF; Size: 100 μg

<span>Each lot of this antibody is quality control tested by </span><a title="View Protocol" href="http://www.biolegend.com/media_assets/support_protocol/BioLegend_IF_protocol.pdf" target="_blank">immunofluorescence staining</a>. For immunofluorescent staining applications, a concentration range of 1-4 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Tubulin-alpha
Clonality: Monoclonal
Clone: 10D8
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: Mouse IgM, κ
Reactivity: Human;Mouse;Rat;All Species




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg 627906 CA627906-BL Each (100µG) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

25 µg 627905 CA627905-BL Each (25µG) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol TUBA
Antigen name Tubulin alpha
Reactivity Human
Conjugation Alexa Fluor® 488
Clonality Monoclonal
Antibody type Primary
Clone
Host Mouse
Isotype IgM kappa
ImmunoFluorescence Yes
Antigen synonyms alpha Tubulin
Gene ID
Concentration
Immunogen
Purification
Form
Storage temperature
Shipping temperature
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty