Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgM Isotype Ctrl Armenian Hamster Monoclonal Antibody (Alexa Fluor® 488) [clone: HTK204]

Supplier: Biolegend
401009
Alexa Fluor® 488 Armenian Hamster IgM Isotype Ctrl [HTK204]; Isotype: Armenian Hamster IgM; Reactivity: Armenian Hamster; Apps: FC, ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control. Use at concentrations comparable to those of the specific antibody of interest.

Type: Control
Antigen: IgM Isotype Ctrl
Clonality: Monoclonal
Clone: HTK204
Conjugation: Alexa Fluor® 488
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgM
Reactivity: Armenian Hamster




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg 401009 CA401009-BL Each (100µG) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol
Antigen name
Reactivity Armenian Hamster
Conjugation Alexa Fluor® 488
Clonality Monoclonal
Antibody type Control
Clone
Host Hamster
Isotype IgM
Flow cytometry Yes
Concentration
Immunogen
Purification
Form
Storage temperature
Shipping temperature
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty