Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG1, k Isotype Ctrl (FC) Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: MOPC-21]

Supplier: Biolegend
400129 400132
Alexa Fluor® 488 Mouse IgG1, κ Isotype Ctrl (FC) [MOPC-21]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC, IF; Size: 25 tests

Each lot of this mouse IgG1, κ isotype control antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, use the isotype control at the same concentration as your primary antibody. Use our <a href="/concentrationlookup" target="_blank">Concentration Lookup</a> tool to find the exact concentrations of your lots of product.

Type: Control
Antigen: IgG1, k Isotype Ctrl (FC)
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Mouse




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 Tests 400129 CA400129-BL Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

25 tests 400132 CA400132-BL Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol
Antigen name
Reactivity Mouse
Conjugation Alexa Fluor® 488
Clonality Monoclonal
Antibody type Control
Clone MOPC-21
Host Mouse
Isotype IgG1 κ
ImmunoFluorescence Yes
Flow cytometry Yes
Concentration
Purification
Form
Storage temperature
Shipping temperature
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping