Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Global Web is currently experiencing issues, technical teams are working to correct the problem.

Anti-IgG2a, k Isotype Ctrl Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: RTK2758]

Supplier: Biolegend
Alexa Fluor® 488 Rat IgG2a, κ Isotype Ctrl [RTK2758]; Isotype: Rat IgG2a, κ; Reactivity: KLH; Apps: FC, ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control. Use at concentrations comparable to those of the specific antibody of interest.

Type: Control
Antigen: IgG2a, k Isotype Ctrl
Clonality: Monoclonal
Clone: RTK2758
Conjugation: Alexa Fluor® 488
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: KLH

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg 400525 CA400525-BL Each (100µG) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol
Antigen name
Conjugation Alexa Fluor® 488
Clonality Monoclonal
Antibody type Control
Clone RTK2758
Host Rat
Isotype IgG2a κ
Flow cytometry Yes
Storage temperature
Shipping temperature
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty