Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Histone H3 Phospho (Ser28) Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: HTA28]

Supplier: Biolegend
641006 641005
Alexa Fluor® 647 anti-Histone H3 Phospho (Ser28) [HTA28]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent intracellular staining with flow cytometric analysis. Please follow the <a title="protocol" href="http://www.biolegend.com/media_assets/support_protocol/BioLegend_EthFIXPERM_032912.pdf" target="_blank">Cell Fixation and Permeabilization Protocol Using 70% Ethanol.</a> For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Histone H3 Phospho (Ser28)
Clonality: Monoclonal
Clone: HTA28
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 Tests 641006 CA641006-BL Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

25 tests 641005 CA641005-BL Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol H3
Antigen name Histone H3
Reactivity Human
Conjugation Alexa Fluor® 647
Clonality Monoclonal
Antibody type Primary
Clone
Host Rat
Isotype IgG2a kappa
Flow cytometry Yes
Antigen synonyms H3FH, H3FT, MGC126886, H3T, HIST1H3C, HIST1H3B, H3FB, HIST1H3J, H3FA, H3FI, H3FD, H3FJ, HIST3H3, HIST1H3E, HIST1H3G, HIST1H3D, HIST1H3I, H3FF, HIST1H3H, HIST1H3F, HIST1H3A, H3FK, H3FL, H3FC HIST1H3E, H3FT, HIST1H3F, H3FK, HIST1H3H, HIST1H3D, H3FH, H3T, H3FD, HIST1H3I, HIST1H3J, H3FC, H3FF, H3FJ, HIST3H3, HIST1H3B, HIST1H3A, H3FI, H3FL, HIST1H3G, MGC126886, H3FB, H3FA, HIST1H3C
Gene ID
Modification phospho-Ser28
Concentration
Immunogen
Purification
Form
Storage temperature
Shipping temperature
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping